Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1720

Supelco

Clomipramine hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Clomipramine hydrochloride, 3-Chloro-10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, Anafranil hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23ClN2 · HCl
CAS Number:
Molecular Weight:
351.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP
traceable to Ph. Eur. C2360000
traceable to USP 1140427

API family

clomipramine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.CN(C)CCCN1c2ccccc2CCc3ccc(Cl)cc13

InChI

1S/C19H23ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-7,10-11,14H,5,8-9,12-13H2,1-2H3;1H

InChI key

WIMWMKZEIBHDTH-UHFFFAOYSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Clomipramine hydrochloride is one of the dibenzazepine tricyclic antidepressants that inhibits noradrenalin and serotonin reuptake in central nervous system. It also shows antimuscarinic properties.[1]

Application

Clomipramine hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in traditional Chinese medicine[2] and pharmaceutical formulations[3][1] by analytical[2][1] and cyclodextrin-modified capillary zone electrophoresis techniques.[3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3902 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Central nervous system,Cardiovascular

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method for simultaneous determination of eight cyclic antidepressants by cyclodextrin-modified capillary zone electrophoresis: applications in pharmaceuticals.
Kou HS, et al.
Analytica Chimica Acta, 525(1), 23-30 (2004)
Multicommutated flow system for the chemiluminometric determination of clomipramine in pharmaceutical preparations.
Marques KL, et al.
Analytica Chimica Acta, 518(1-2), 31-36 (2004)
HPLC and GC?MS screening of Chinese proprietary medicine for undeclared therapeutic substances.
Liu SY, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(5-6), 983-992 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service