Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1592

Supelco

Tenofovir

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid monohydrate, 1-(6-Aminopurin-9-yl) propan-2-yl-oxymethylphosphonic acid, TEN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N5O4P · H2O
CAS Number:
Molecular Weight:
305.23
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1643601

API family

tenofovir

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[P](=O)(O)(O)CO[C@@H](C[n]1c2ncnc(c2nc1)N)C.O

InChI

1S/C9H14N5O4P.H2O/c1-6(18-5-19(15,16)17)2-14-4-13-7-8(10)11-3-12-9(7)14;/h3-4,6H,2,5H2,1H3,(H2,10,11,12)(H2,15,16,17);1H2/t6-;/m1./s1

InChI key

PINIEAOMWQJGBW-FYZOBXCZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Tenofovir is a nucleotide analog reverse transcriptase inhibitor, and is widely used as an antiretroviral agent, for the treatment of HIV infection.

Application

Tenofovir may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance thin layer chromatography (HPTLC) technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3475 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPTLC method for the simultaneous estimation of emtricitabine and tenofovir in tablet dosage form
Joshi M, et al.
Indian Journal of Pharmaceutical Sciences, 71(1), 95-95 (2009)
Validated HPTLC method for the determination of tenofovir as bulk drug and in pharmaceutical dosage form
Pradeep K, et al.
Research Journal of Chemical Sciences, 1(7), 33-37 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service