Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1490

Supelco

Monoethanolamine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ethanolamine, 2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
Beilstein:
505944
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001184
traceable to USP 1445925

vapor density

2.1 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

description

Pharmaceutical Secondary Standard; Certified Reference Material

API family

ethanolamine

CofA

current certificate can be downloaded

autoignition temp.

1436 °F

expl. lim.

17 %

packaging

pkg of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.454 (lit.)

bp

170 °C (lit.)
69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Monoethanolamine is majorly used in pharmaceutical formulations in the buffer systems, and emulsion preparations. It can also be used as a solvent for fats and oils and to synthesize many salts having therapeutic essence.[1]

Application

Monoethanolamine may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations and cosmetic products by chromatography.[2][3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2076 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous analysis of mono-, di-, and tri-ethanolamine in cosmetic products using liquid chromatography coupled tandem mass spectrometry
Shin KO and Lee YM
Archives of Pharmacal Research, 39(1), 66-72 (2016)
Handbook of Pharmaceutical Excipients (2009)
Monoethanolamine
USP42-NF37
United States Pharmacopeia/National Formulary, 31(5), 5836-5836 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service