Skip to Content
MilliporeSigma

1.00844

Ethanolamine

EMPROVE® ESSENTIAL, Ph. Eur., BP, NF

Synonym(s):

Ethanolamine, 2-Aminoethanol, Monoethanolamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOCH2CH2NH2
CAS Number:
Molecular Weight:
61.08
UNSPSC Code:
12191502
EC Index Number:
205-483-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, NF, Ph. Eur.

vapor pressure

0.5 hPa ( 20 °C)

description

EMPROVE® ESSENTIAL BP,NF

product line

EMPROVE® ESSENTIAL

assay

98.0-100.5% (acidimetric)

form

liquid

autoignition temp.

410 °C

expl. lim.

3.4-27 % (v/v)

technique(s)

pH adjustment: suitable

pH

12.1 (20 °C, 100 g/L in H2O)

kinematic viscosity

20 cSt(23 °C)

bp

171 °C/1013 hPa

mp

10.5 °C

transition temp

flash point 92.5 °C

density

1.02 g/cm3 at 20 °C

application(s)

liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation

storage temp.

15-25°C

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ethanolamine, a primary amine, is typically used as a buffering agent in small molecule liquid formulations. It may also be used in large molecule downstream purification as a chromatography buffer component.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Ethanolamine


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point (°F)

195.8 °F - Pensky-Martens closed cup

Flash Point (°C)

91 °C - Pensky-Martens closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.