Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1442

Supelco

Flunixin Meglumine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Flunixin meglumine, 2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid meglumine salt, Banamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11F3N2O2·C7H17NO5
CAS Number:
Molecular Weight:
491.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000443
traceable to USP 1274607

API family

flunixin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.Cc1c(Nc2ncccc2C(O)=O)cccc1C(F)(F)F

InChI

1S/C14H11F3N2O2.C7H17NO5/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12;1-8-2-4(10)6(12)7(13)5(11)3-9/h2-7H,1H3,(H,18,19)(H,20,21);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1

InChI key

MGCCHNLNRBULBU-WZTVWXICSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flunixin meglumine is a nonsteroidal anti-inflammatory drug and a potent cyclo-oxygenase (COX) inhibitor. It is commonly used as an analgesic and antipyretic in animals.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Flunixin meglumine (IC50 = 1 nM) can be used as a drug for animals for the management of intestinal ischaemia, colic, and endotoxemia in horses.
Flunixin meglumine may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by spectrophotometry and high performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

COX1, COX2 and PLA2 inhibitor non-narcotic analgesic, anti-pyretic, anti-inflammatory.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2750 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flunixin Meglumine
USP42/NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 39(4), 1888-1888 (2014)
Micellar high performance liquid chromatographic determination of flunixin meglumine in bulk, pharmaceutical dosage forms, bovine liver and kidney
Belal FF, et al.
Analytical Chemistry Research, 3, 63-69 (2015)
Chapter 3.6.3 Flunixin
Textbook of Rabbit Medicine, 137-177 (2014)
Spectrophotometric Methods for Simultaneous Determination of Oxytetracycline HCl and Flunixin Meglumine in Their Veterinary Pharmaceutical Formulation
Merey HA, et al.
Journal of Analytical Methods in Chemistry, 2017 (2017)
Luiz A M Penteado et al.
Journal of oral science, 55(3), 225-232 (2013-09-18)
Bone regeneration is an important objective in clinical dental practice and has been used for different applications. The aim of this study was to evaluate the effectiveness of platelet-rich plasma (PRP) and bioactive glass (BG) for bone healing of surgical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service