Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1253

Supelco

Acarbose

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Acarbose, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H43NO18
CAS Number:
Molecular Weight:
645.60
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000500
traceable to USP 1000521

API family

acarbose

CofA

current certificate can be downloaded

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@@H]1N[C@H]4C=C(CO)[C@@H](O)[C@H](O)[C@H]4O

InChI

1S/C25H43NO18/c1-6-11(26-8-2-7(3-27)12(30)15(33)13(8)31)14(32)19(37)24(40-6)43-22-10(5-29)42-25(20(38)17(22)35)44-21-9(4-28)41-23(39)18(36)16(21)34/h2,6,8-39H,3-5H2,1H3/t6-,8+,9-,10-,11-,12-,13+,14+,15+,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-/m1/s1

InChI key

XUFXOAAUWZOOIT-SXARVLRPSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Acarbose is an oral antidiabetic drug, a glucosidase inhibitor used in the management of non-insulin-dependent diabetes mellitus (NIDDM).

Application

Acarbose may be used as a pharmaceutical reference standard in determining the concentration of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Modified tetrasaccharide that acts as a reversible α -glucosidase inhibitor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9057 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of liquid chromatography and capillary electrophoresis methods for acarbose determination in pharmaceutical tablets.
Cherkaoui S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 18(4-5), 729-735 (1998)
Analysis of the antidiabetic drug acarbose by capillary electrophoresis.
Rethfeld I and Blaschke G
Journal of Chromatography. B, Biomedical Sciences and Applications, 700(1-2), 249-253 (1997)
Patrick Schwientek et al.
BMC genomics, 13, 112-112 (2012-03-27)
Actinoplanes sp. SE50/110 is known as the wild type producer of the alpha-glucosidase inhibitor acarbose, a potent drug used worldwide in the treatment of type-2 diabetes mellitus. As the incidence of diabetes is rapidly rising worldwide, an ever increasing demand
High performance liquid chromatography of acarbose and its metabolite on porous graphitic carbon column.
Daali Y, et al.
Journal of Separation Science, 25(5-6), 280-284 (2002)
Gary Williamson
Molecular nutrition & food research, 57(1), 48-57 (2012-11-28)
Excessive post-prandial glucose excursions are a risk factor for developing diabetes, associated with impaired glucose tolerance. One way to limit the excursion is to inhibit the activity of digestive enzymes for glucose production and of the transporters responsible for glucose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service