Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C0650000

Cefadroxil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cefadroxil monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O5S · H2O
CAS Number:
Molecular Weight:
381.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefadroxil

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

S1[C@H]2N(C(=C(C1)C)C(=O)O)C(=O)[C@H]2NC(=O)[C@H](N)c3ccc(cc3)O.O

InChI

1S/C16H17N3O5S.H2O/c1-7-6-25-15-11(14(22)19(15)12(7)16(23)24)18-13(21)10(17)8-2-4-9(20)5-3-8;/h2-5,10-11,15,20H,6,17H2,1H3,(H,18,21)(H,23,24);1H2/t10-,11-,15-;/m1./s1

InChI key

NBFNMSULHIODTC-CYJZLJNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefadroxil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Marta Veses-Garcia et al.
Frontiers in microbiology, 9, 1530-1530 (2018-07-26)
Achieving fast antimicrobial susceptibility results is a primary goal in the fight against antimicrobial resistance. Standard antibiotic susceptibility testing (AST) takes, however, at least a day from patient sample to susceptibility profile. Here, we developed and clinically validated a rapid
Fatmanur Tuğcu-Demiröz et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 62, 105-110 (2014-06-03)
The aim of the present study was to develop a method for water flux reabsorption measurement in Doluisio's Perfusion Technique based on the use of phenol red as a non-absorbable marker and to validate it by comparison with gravimetric procedure.
Michael Zakrewsky et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(37), 13313-13318 (2014-08-27)
Biofilm-protected microbial infections in skin are a serious health risk that remains to be adequately addressed. The lack of progress in developing effective treatment strategies is largely due to the transport barriers posed by the stratum corneum of the skin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service