Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL82649

Scopolin

phyproof® Reference Substance

Synonym(s):

7-Hydroxy 6-methoxycoumarin 7-glucoside, Scopoletin 7-glucoside, Scopoloside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2cc3c(cc2OC)CCC(=O)O3

InChI

1S/C16H20O9/c1-22-9-4-7-2-3-12(18)23-8(7)5-10(9)24-16-15(21)14(20)13(19)11(6-17)25-16/h4-5,11,13-17,19-21H,2-3,6H2,1H3/t11-,13-,14+,15-,16-/m1/s1

InChI key

WSHSQJSPOLYHLM-YMILTQATSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: hedera

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafaliya Rutul V et al.
Phytochemistry, 182, 112594-112594 (2020-12-20)
The scopoletin one of the major bioactive components of Convolvulus prostratus Forssk known to have a role in acetylcholinesterase inhibitor, memory enhancer, antimicrobial, antioxidative etc. properties are investigated in the present study. The concentration of scopoletin in C. prostratus is
Kevin Robe et al.
The New phytologist, 229(4), 2062-2079 (2020-11-19)
Iron (Fe) is a major micronutrient and is required for plant growth and development. Nongrass species have evolved a reduction-based strategy to solubilize and take up Fe. The secretion of Fe-mobilizing coumarins (e.g. fraxetin, esculetin and sideretin) by plant roots
Christopher J Harbort et al.
Cell host & microbe, 28(6), 825-837 (2020-10-08)
Plants benefit from associations with a diverse community of root-colonizing microbes. Deciphering the mechanisms underpinning these beneficial services are of interest for improving plant productivity. We report a plant-beneficial interaction between Arabidopsis thaliana and the root microbiota under iron deprivation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service