Skip to Content
MilliporeSigma
All Photos(1)

Documents

P0521

Millipore

Peptone from Glycine max (soybean)

Type IV, powder, Component of microbiological growth media

Synonym(s):

Protein from soybean, Soya peptone type IV

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106212
NACRES:
NA.85

biological source

Glycine max (soybean)

Quality Level

sterility

non-sterile

type

Type IV

form

powder

nitrogen analysis

~2.0% amino, ~8.0% total

application(s)

food and beverages
microbiology
vitamins, nutraceuticals, and natural products

storage temp.

10-30°C

General description

Peptone from Glycine max (soybean), Type IV is an enzymatically hydrolysed soy protein. This peptone is used in fermentation broths as a carbohydrate and nitrogen source (peptides and free amino acids), producing consistent results. It acts comparatively well as peptones obtained from animal sources.

Application

Peptone acts as the main source of nitrogen in microbiological culture media to meet nutrient requirements of microorganisms being cultivated.
Peptone from Glycine max as a nutrient, displays comparative or better growth features of cultivated microorganisms than peptone from other sources

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meiyappan Lakshmanan et al.
Biotechnology and bioengineering, 116(9), 2117-2129 (2019-05-09)
Chinese hamster ovary (CHO) cells are the most prevalent mammalian cell factories for producing recombinant therapeutic proteins due to their ability to synthesize human-like post-translational modifications and ease of maintenance in suspension cultures. Currently, a wide variety of CHO host

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service