Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O0180000

Orciprenaline sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Metaproterenol hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H17NO3 · 1/2H2SO4
CAS Number:
Molecular Weight:
260.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

orciprenaline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CC(C)NCC(O)c1cc(O)cc(O)c1.CC(C)NCC(O)c2cc(O)cc(O)c2

InChI

1S/2C11H17NO3.H2O4S/c2*1-7(2)12-6-11(15)8-3-9(13)5-10(14)4-8;1-5(2,3)4/h2*3-5,7,11-15H,6H2,1-2H3;(H2,1,2,3,4)

InChI key

MKFFGUZYVNDHIH-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Orciprenaline sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Codruţa Cobzac et al.
Journal of chromatographic science, 52(9), 1095-1103 (2013-10-29)
The retention behavior for a series of amine neurotransmitters, their precursors, metabolites and structurally related drugs has been investigated in reversed-phase thin-layer chromatography using RP-18, RP-8, RP-2, CN and Diol stationary phases and mixtures of phosphate buffer and methanol as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service