Skip to Content
MilliporeSigma
All Photos(1)

Documents

B0477000

Benserazide hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

DL-Serine 2-(2,3,4-trihydroxybenzyl)hydrazide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N3O5 · HCl
CAS Number:
Molecular Weight:
293.70
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benserazide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.NC(CO)C(=O)NNCc1ccc(O)c(O)c1O

InChI

1S/C10H15N3O5.ClH/c11-6(4-14)10(18)13-12-3-5-1-2-7(15)9(17)8(5)16;/h1-2,6,12,14-17H,3-4,11H2,(H,13,18);1H

InChI key

ULFCBIUXQQYDEI-UHFFFAOYSA-N

Gene Information

human ... DDC(1644)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benserazide hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Inhibitor of L-aromatic amino acid decarboxylase.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wai Kin D Ko et al.
Neurobiology of disease, 70, 138-148 (2014-06-28)
Regulators of G-protein signalling (RGS) proteins are implicated in striatal G-protein coupled receptor (GPCR) sensitisation in the pathophysiology of l-DOPA-induced abnormal involuntary movements (AIMs), also known as dyskinesia (LID), in Parkinson's disease (PD). In this study, we investigated RGS protein
Jacob Wienecke et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(36), 11984-12000 (2014-09-05)
Serotonin (5-HT), an important modulator of both sensory and motor functions in the mammalian spinal cord, originates mainly in the raphe nuclei of the brainstem. However, following complete transection of the spinal cord, small amounts of 5-HT remain detectable below
Sylvia Navailles et al.
CNS neuroscience & therapeutics, 20(7), 671-678 (2014-04-30)
Serotonin (5-HT) neurons mediate the ectopic release of dopamine (DA) induced by L-DOPA in the Parkinsonian brain. We hypothesized that the participation of noradrenalin transporters (NET) in the clearance of DA may account for the lower effect of L-DOPA in
Dalila Mango et al.
Experimental neurology, 261, 733-743 (2014-09-01)
γ-Aminobutyric acid A receptor (GABAAR)-mediated postsynaptic currents were recorded in brain slices from substantia nigra pars reticulate neurons. The selective adenosine A1 receptor (A1R) antagonist, 8-cyclopentyl-1,3-dipropylxanthine (DPCPX), increased the frequency, but not the amplitude of spontaneous inhibitory post-synaptic currents (IPSCs)
Yong Wang et al.
Synapse (New York, N.Y.), 68(8), 332-343 (2014-03-07)
l-3,4-Dihydroxyphenylalanine (L-DOPA) is the most widely used drug for the treatment of Parkinson's disease. Unfortunately, chronic administration of this dopamine precursor causes L-DOPA-induced dyskinesia (LID), which is a debilitating complication whose pathogenesis remains unclear. In this study, we compared gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service