Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST762

Magnetic moment standard - nickel disk

NIST® SRM® 762

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

disc

packaging

pkg of 6 mm dia

manufacturer/tradename

NIST®

application(s)

general analytical

General description

This Standard Reference Material (SRM) is intended for use in the calibration of magnetometers (such as vibrating sample magnetometers) used in the measurement of the magnetic properties of materials. SRM 762 consists of a nickel disk nominally 6 mm in diameter with a thickness of 0.127 mm and a mass of 32 mg ± 1 mg. For more information, please refer to the COA and SDS.

SRM 762_cert

SRM 762_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Nickel(Ni)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service