Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST2133

Phosphorus implant in silicon depth profile standard

NIST® SRM® 2133

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of each

manufacturer/tradename

NIST®

application(s)

semiconductor

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in calibrating secondary ion response to minor and trace levels of phosphorus in a silicon matrix by the analytical technique of secondary ion mass spectrometry (SIMS). SRM 2133 is intended for calibrating the response of a SIMS instrument for phosphorus in a silicon matrix under a specific set of instrumental conditions. It may also be used by a laboratory as a transfer standard for the calibration of working standards of phosphorus in silicon. This SRM consists of a 1 cm × 1 cm single crystal silicon substrate that has been ion-implanted with the isotope 31P at a nominal energy of 100 keV. For more information, please refer to the COA and SDS.

SRM 2133_cert

SRM 2133_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Phosphorus (31P)

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service