Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST696

Bauxite

NIST® SRM® 696, Surinam

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 60 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is in th eform of fine powder (nominal particle size < 0.08 mm) and is intended for use in evaluating chemical and instrumental methods of analysis.

SRM 696_cert

SRM 696_SDS

Application


  • Epidural Nalbuphine in Orthopedic Surgeries: Nalbuphine hydrochloride was evaluated as an epidural adjuvant to bupivacaine for postoperative analgesia in lower limb orthopedic surgeries. This study underscores its effective role in enhancing pain management protocols, demonstrating significant improvements in patient comfort during the postoperative phase (Farmawy et al., 2023).

  • Pharmacokinetic Study of Nalbuphine Nasal Spray: Comparative pharmacokinetics of nalbuphine nasal spray versus injection solution in healthy volunteers were studied, providing valuable data on the bioavailability and systemic effects of different administration routes. This research aids in developing patient-friendly nalbuphine formulations for pain management (Tymko et al., 2023).

  • Pharmacokinetic Profiling in Anesthesia: A validated ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) method was developed for determining nalbuphine in human plasma. This method was applied in a pharmacokinetic study of patients undergoing general anesthesia, enhancing the understanding of its pharmacological behavior during surgical procedures (Gao et al., 2023).


Other Notes

Certified for the analytes listed below.
Aluminium oxide (AL2O3), Iron oxide (Fe2O3), Silicon dioxide (SiO2), Titanium dioxide (TiO2), Zirconium dioxide (ZrO2), Phosphorus pentoxide (P2O5), Vanadium pentoxide (V2O5), Chromium oxide (Cr2O3), Calcium oxide (CaO), Mangnesium oxide (Mgo), Manganese oxide (MnO), Zinc oxide (Zno), Potassium oxide (K2O), Sulfur trioxide (SO3)

Reference/Informational value is provided for the following analytes.
Barium oxide (BaO), Sodium oxide (Na2O), Cerium (Ce), Cobalt (Co), Hafnium (Hf), Scandium (Sc)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service