Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST3950

Vitamin B6 in frozen human serum

NIST® SRM® 3950

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

packaging

pkg of 2 vials, 1 level ea

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

storage temp.

−70°C

General description

Pyridoxal 5′-phosphate (PLP) is the major circulating form of vitamin B6 and the most common direct measure of this vitamin in serum or plasma. A unit of the vitamin B6 standard reference material (SRM) 3950 consists of two stoppered vials of frozen human serum, one vial each at two different concentration levels. Each vial contains 1.0 mL of human serum.

SRM 3950_cert

SRM 3950 _SDS

Application

Vitamin B6 in frozen human serum standard reference material (SRM) 3950 is intended primarily for use in evaluating the accuracy of procedures for the determination of the vitamin B6 metabolite pyridoxal 5′-phosphate (PLP) in human serum. It is also intended for use in validating working or secondary reference materials.

Features and Benefits

The use of this material will provide confidence in the assignment of values to the in-house assay calibrators.

Preparation Note

  • SRM 3950 is provided as a frozen serum that should be allowed to thaw at room temperature (20 °C to 25 °C) under subdued light.
  • After the material is thawed, it should be used immediately.
  • The contents of the vial should then be gently mixed prior to the removal of a test portion for analysis.
  • Precautions should be taken to expose serum only under subdued lighting conditions.

Other Notes

  • SRM 3950 is intended for research use.
  • Details on expiration, storage, safety, usage, and source are provided in the NIST certificate
  • Information on biomaterials, disposal, and transport is available in the SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaochun Zhang et al.
Practical laboratory medicine, 21, e00160-e00160 (2020-04-30)
Vitamin B6 deficiency is associated with a wide spectrum of clinical syndromes. While vitamin B6 status is primarily assessed by measuring the biologically active form of the vitamin, pyridoxal 5-phosphate (PLP), concurrent measurement of the final metabolite 4-pyridoxic acid (PA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service