Skip to Content
MilliporeSigma
All Photos(1)

Documents

731188

Sigma-Aldrich

Thiamine-(4-methyl-13C-thiazol-5-yl-13C3) hydrochloride

99 atom % 13C, 98% (CP)

Synonym(s):

Vitamin B1(4-methyl-13C-thiazol-5-yl-13C3) hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C8H17ClN4OS · HCl
CAS Number:
Molecular Weight:
341.24
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white

mp

250 °C (dec) (lit.)

mass shift

M+4

storage temp.

−20°C

SMILES string

Cl.[Cl-].Cc1ncc(C[n+]2[13cH]s[13c](CCO)[13c]2[13CH3])c(N)n1

InChI

1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1/i1+1,7+1,8+1,11+1;;

InChI key

DPJRMOMPQZCRJU-VJFONHOHSA-M

Application

Thiamine-(4-methyl-13C-thiazol-5-yl-13C3) hydrochloride is also known as vitamin B1(4-methyl-13C-thiazol-5-yl-13C3) hydrochloride. It can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples by mass spectrometry (MS).

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rohiverth Guarecuco et al.
Science advances, 6(41) (2020-10-11)
Tumor environment influences anticancer therapy response but which extracellular nutrients affect drug sensitivity is largely unknown. Using functional genomics, we determine modifiers of l-asparaginase (ASNase) response and identify thiamine pyrophosphate kinase 1 as a metabolic dependency under ASNase treatment. While

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service