Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2242A

Relative Intensity Correction Standard for Raman Spectroscopy: 532 nm Excitation

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 1 ea (1 g)

manufacturer/tradename

NIST®

application(s)

general analytical

storage temp.

15-25°C

General description

This Standard Reference Material (SRM) is a certified spectroscopic standard for the correction of the relative intensity of Raman spectra obtained with instruments employing 532 nm laser excitation. A unit of SRM 2242aconsists of an optical glass slide that emits a broadband luminescence spectrum when excited at 532 nm. This SRMis approximately 10 mm in width × 10 mm in length × 1.65 mm in thickness, with both surfaces optically polished, and furnished with two mounts.

SRM 2242A_cert

SRM 2242A_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service