Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST185I

Potassium hydrogen phthalate

NIST® SRM® 185i, pH standard

Synonym(s):

Potassium phthalate monobasic, KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for SM 2310
suitable for SM 2320

form

crystals

packaging

pkg of 60 g

manufacturer/tradename

NIST®

mp

295-300 °C (dec.) (lit.)

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

SRM 185I is certified as a pH standard [pH(S)].   

Each unit of SRM 185I consists of 60 g of potassium hydrogen phthalate.

SRM 185I _SDS

SRM 185I _CoA

Application

Potassium hydrogen phthalate standard reference material (SRM) is intended for use in preparing solutions for calibrating electrodes for pH measuring systems.

Features and Benefits

  • SRM 185I is available with certified pH(S) values and their expanded uncertainties. 
  • Certified values are derived from potential measurements of cells without liquid junction by the primary measurement method. 
  • Certified values are metrologically traceable to the International System of Units (SI) of amount-of-substance and mass and to the definition of single ion activity. 
  • The NIST certificate is provided with maintenance, storage, handling, and use instructions.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service