Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1450E

Thermal Conductivity - Fibrous Glass Board

pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 1 ea

manufacturer/tradename

NIST® SRM 1450e

General description

This Standard Reference Material (SRM) is intended primarily for use in the measurement of the thermal conductivity or thermal resistance of insulation materials. SRM 1450e is a high-density fibrous glass board certified for bulk density and thermal conductivity. The SRM can be used in conjunction with ASTM C177 or ASTM C518. A unit of SRM 1450e consists of a square panel of fibrous glass and phenolic binder molded into a semi-rigid board. The nominal dimensions of a unit are 610 mm × 610 mm × 25 mm.

SRM 1450E_cert

SRM 1450E_SDS

Application


  • High-temperature insulation using fibrous glass board: The study by Carvajal SA, Garboczi EJ, and Zarr RR utilized fibrous glass board as a reference material for thermal resistance measurements (Carvajal et al., 2019).

  • Thermal conductivity standards for fibrous glass board: Zarr RR, Heckert NA, and Leigh SD conducted a retrospective analysis of the Standard Reference Material 1450, fibrous glass board, establishing its thermal insulation measurement standards that support research applications in biochemistry and biotechnological fields (Zarr et al., 2014).

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service