Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1240C

Aluminum alloy 3004

NIST® SRM® 1240c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1240C contains a disk with a diameter of ~6.3 cm and a thickness of 19 mm.


SRM 1240C_cert


SRM 1240C_SDS

Application

Aluminum alloy 3004 SRM is intended for the assessment of instrumental methods of analysis, such as glow discharge optical emission spectrometry, spark source optical emission spectrometry, and X-ray fluorescence spectrometry.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction values, expanded uncertainty coverage factor of elements in aluminum alloy.
  • The reference and information values of elements of aluminum alloy are presented as mass fractions.
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.
  • The certified values are metrologically traceable to the SI unit of mass, expressed as a percent.

Other Notes

Certified for the analytes listed below.
Silicon (Si), Iron (Fe), Copper (Cu), Manganese (Mn), Magnesium (Mg), Nickel (Ni), Zinc (Zn), Titanium (Ti), Vanadium (V), Gallium (Ga)

Reference/Informational value is provided for the following analytes.
Beryllium (Be), Cadmium (Cd), Chromium (Cr), Tin (Sn), Zirconium (Zr), Strontium (Sr), Lead (Pb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service