Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1258-I

Aluminum alloy 6011

NIST® SRM® 1258-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 1258-I contains a wrought and annealed disk with dimensions 35 mm in diameter and 19 mm in thickness.



SRM 1258-I_cert

SRM 1258-I_SDS

Application

Aluminum Alloy 6011 standard reference material (SRM) is primarily intended for evaluating the methods used to analyze elements in aluminum alloys of similar matrix.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified and reference mass fraction values, coverage factor (k),and expanded uncertainty for total elements in an aluminum matrix of SRM1258-I.
  • Information values for beryllium and gallium are reported as mass fractions.
  • The certified values are metrologically traceable to the derived SI unit of mass fraction, expressed in%.
  • The NIST certificate is provided with expiration certificate, storage, handling, maintenance,and use instructions.

Other Notes

Certified for the analytes listed below.
Silicon (Si), Iron (Fe), Copper (Cu), Manganese (Mn), Magnesium (Mg), Zinc (Zn)

Reference/Informational value is provided for the following analytes.
Beryllium (Be), Gallium (Ga), Chromium (Cr), Nickel (Ni), Titanium (Ti), Beryllium (Be), Gallium (Ga)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service