Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2392

Supelco

Meprobamate

analytical standard

Synonym(s):

2-Methyl-2-propylpropane-1,3-diol dicarbamate, MPB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O4
CAS Number:
Molecular Weight:
218.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CCCC(C)(COC(N)=O)COC(N)=O

InChI

1S/C9H18N2O4/c1-3-4-9(2,5-14-7(10)12)6-15-8(11)13/h3-6H2,1-2H3,(H2,10,12)(H2,11,13)

InChI key

NPPQSCRMBWNHMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Enhances GABAA receptor activity by increasing Cl currents. At high concentrations, blocks NMDA glutamate receptor currents.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arun Kumar et al.
Regulatory toxicology and pharmacology : RTP, 57(2-3), 146-156 (2010-02-16)
This study presents a step-wise development of a quantitative pharmaceutical risk assessment (QPhRA, hereafter) framework, including Monte Carlo uncertainty analysis for meprobamate, carbamazepine, and phenytoin during (1) accidental exposures of stream water and fish consumption and (2) direct ingestion of
Shane A Snyder et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 61(1), 145-154 (2010-01-09)
The presence of pharmaceuticals and endocrine disrupting compounds (EDCs) in the environment raises many questions about risk to the environment and risk to human health. Researchers have attributed adverse ecological effect effects to the presence of these compounds, particularly EDCs
Fabien Bévalot et al.
International journal of legal medicine, 125(3), 463-468 (2011-02-22)
The use of vitreous humor (VH) as an alternative matrix to blood in the field of forensic toxicology has been described for numerous drugs. Interpretation of drug concentrations measured in VH, as in other matrices, requires statistical analysis of a
Iain M McIntyre et al.
Journal of analytical toxicology, 36(3), 177-181 (2012-03-16)
Carisoprodol is a therapeutic and occasionally abused centrally acting muscle relaxant. We compare central blood and liver concentrations of carisoprodol and the metabolite meprobamate to concentrations in peripheral blood in 11 medical examiner cases. Specimens were initially screened for alcohol
Gudrun Høiseth et al.
European journal of clinical pharmacology, 68(11), 1561-1565 (2012-04-25)
Carisoprodol, a frequently used muscle relaxant, can cause potentially fatal intoxications. Conversion to its active metabolite meprobamate is almost solely mediated by cytochrome P450 2C19 (CYP2C19), and mutations in this enzyme could have significant effects on serum concentrations. The objective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service