Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

I0890

Sigma-Aldrich

Immersion oil

viscosity 1,250 cSt (lit.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

biological source

synthetic

Quality Level

viscosity

1,250 cSt(lit.)

application(s)

hematology
histology

storage temp.

room temp

General description

Immersion oil has a larger impact on 3D resolution. Specifically, matching the immersion oil refractive index to glass, reliably enhances the ability to visualize dendritic spines.

Application

Immersion oil has been used in epifluorescent microscopy and cell identification.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphological controls on cannibalism in a planktonic marine phagotroph
Martel, et al.
Protist, 159(1), 41-51 (2008)
Impact of immersion oils and mounting media on the confocal imaging of dendritic spines.
Peterson BM, et al.
Journal of Neuroscience Methods, 242, 106-111 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service