Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1517

Supelco

Fenoprofen calcium salt hydrate

analytical standard, for drug analysis

Synonym(s):

(±)-2-(3-Phenoxyphenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C30H26O6Ca
CAS Number:
Molecular Weight:
522.60
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

Assay

≥97% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O.[Ca].CC(C(O)=O)c1cccc(Oc2ccccc2)c1

InChI

1S/C15H14O3.Ca.H2O.2H/c1-11(15(16)17)12-6-5-9-14(10-12)18-13-7-3-2-4-8-13;;;;/h2-11H,1H3,(H,16,17);;1H2;;

InChI key

JWCQUXFJNIIYQZ-UHFFFAOYSA-N

Application

Fenoprofen calcium salt hydrate may be used as an analytical reference standard for the quantification of the analyte in environmental samples using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Multiresidue analysis of acidic and polar organic contaminants in water samples by stir-bar sorptive extraction?liquid desorption?gas chromatography?mass spectrometry
Quintana BJ, et al.
Journal of Chromatography A, 1174(1-2), 27-39 (2007)
Frederik André Hansen et al.
Analytica chimica acta, 1143, 109-116 (2021-01-02)
In this work, we investigated for the first time hydrophobic deep eutectic solvents (DES) as supported liquid membrane (SLM) for electromembrane extraction (EME). Camphor, coumarin, DL-menthol, and thymol were used as non-ionic DES components. Different DESs compositions were tested, to
Josiane Cristófani Poggi et al.
Journal of clinical pharmacology, 46(11), 1328-1336 (2006-10-20)
The objective of the present study was to assess the influence of type 1 and type 2 diabetes mellitus on the enantioselective pharmacodynamics and pharmacokinetics of fenoprofen. Patients with diabetes mellitus type 1 (n = 7) or type 2 (n
Zhaoyan Wang et al.
Journal of chromatography. A, 1218(13), 1775-1779 (2011-02-24)
A highly sensitive method for enantioseparation of trace fenoprofen and amino acid derivatives by capillary electrophoresis (CE) with vancomycin as the chiral selector was developed. Several CE techniques, such as the partial filling, large-volume sample stacking with EOF as pump
Cynthia L Stevenson et al.
Journal of pharmaceutical sciences, 94(9), 1861-1880 (2005-07-30)
Pharmaceutical solids have generally been characterized as either three-dimensional crystals or amorphous solids based on X-ray powder diffraction and modulated temperature differential scanning calorimetry. In contrast, fewer examples of thermotropic and lyotropic liquid crystals, or mesophases, appear in the pharmaceutical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service