Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMFD066

Corundum Powder (Particle size distribution)

ERM®, certified reference material

Synonym(s):

Corundum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

SMILES string

[Al]21O[Al](O2)O1

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service