Skip to Content
MilliporeSigma
All Photos(1)

Documents

ERMEB530A

Aluminum gold alloy

0.1% Au, 0.1 mm foil, ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

product name

Aluminum gold alloy (0.1% Au), 0.1 mm foil, ERM®, certified reference material

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

format

matrix material

General description

Certified for the gold content in Aluminum.

ERM-EB530A_cert
ERM-EB530_report
ERM-EB530_origin

Analysis Note

For more information please see:
ERMEB530A

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service