Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST915C

Calcium Carbonate

NIST® SRM® 915c

Synonym(s):

Calcium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

packaging

pkg of 20 g

manufacturer/tradename

NIST®

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 915c contains ~ 20 g of material in a single glass container.
For more information, please refer to the SDS and COA.
SRM 915C_cert
SRM 915C_SDS
Application
Calcium Carbonate standard reference material (SRM) is primarily intended for calibrating, validating, and standardizing analytical methods used in clinical analysis for determining calcium (Ca) and carbonate.

Features and Benefits

  • The certified mass fraction values for elements are provided on a dry-mass basis and values are metrologically traceable to the SI unit of mass, expressed as a percentage.
  • Provided with non-certified values ideal for method harmonization, method development, and process control.
  • Expiry date, instructions for storage and usage are detailed on the NIST certificate.

Analysis Note

Standard Reference Material (SRM) 915c Calcium Carbonate is certified as a chemical substance of known purity. It is intended for use in the calibration and standardization of procedures for calcium (Ca) determinations employed in clinical analysis and for routine critical evaluation of the daily working standards used in these procedures. A unit of SRM 915c consists of a single glass bottle containing 20 g of the material.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service