Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMBF421B

EH92-527-1 Potato (level 1)

ERM®, certified reference material, 100% EH92-527-1 potato, for the detection of GMO potato (EH92-527-1) with PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

quality

for the detection of GMO potato (EH92-527-1) with PCR

manufacturer/tradename

JRC

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Analysis Note

For more information please see:
ERMBF421B

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service