Skip to Content
MilliporeSigma
All Photos(1)

Documents

D1410000

Diethyl phthalate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1912500
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

API family

diethyl phthalate

autoignition temp.

854 °F

expl. lim.

0.75 %, 187 °F

manufacturer/tradename

EDQM

refractive index

n20/D 1.502 (lit.)

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diethyl phthalate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Api
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 39(2), 97-108 (2001-03-27)
Diethyl phthalate (DEP; CAS No. 84-66-2) has many industrial uses, as a solvent and vehicle for fragrance and cosmetic ingredients and subsequent skin contact. This review focuses on its safety in use as a solvent and vehicle for fragrance and
Diethyl phthalate: a perspective.
M A Kamrin et al.
Journal of clinical pharmacology, 31(5), 484-489 (1991-05-01)
Antonia M Calafat et al.
Environmental health perspectives, 114(11), 1783-1789 (2006-11-17)
The probability of nonoccupational exposure to phthalates is high given their use in a vast range of consumables, including personal care products (e.g., perfumes, lotions, cosmetics), paints, industrial plastics, and certain medical devices and pharmaceuticals. Phthalates are of high interest
Gurusankar Saravanabhavan et al.
The Science of the total environment, 500-501, 191-198 (2014-09-15)
We have analyzed the trends in the body-weight-adjusted urinary monoethyl phthalate (MEP) concentrations and the diethyl ethyl phthalate (DEP) daily intake estimates in the general Canadian population (aged 6-49 years) using the Canadian Health Measures Survey 2007-2009 dataset. The creatinine
T Fierens et al.
Environment international, 73, 117-127 (2014-08-13)
In this study, a semi-probabilistic modelling approach was applied for the estimation of the long-term human dietary exposure to phthalates--one of world's most used families of plasticisers. Four phthalate compounds were considered: diethyl phthalate (DEP), di-n-butyl phthalate (DnBP), benzylbutyl phthalate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service