Skip to Content
MilliporeSigma

Skip To

41320

Dimethyl phthalate

PESTANAL®, analytical standard

Synonym(s):

DMP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H4-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
205-011-6
Beilstein/REAXYS Number:
1911460
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

assay

≥99.5% (GC)

autoignition temp.

1033 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.515 (lit.), n20/D 1.516

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COC(=O)c1ccccc1C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

General description

Dimethyl phthalate can widely find applications in industries, for the manufacture of polyvinyl chloride and cellulose ester-based plastics like cellulose acetate and cellulose butyrate.
Dimethyl phthalate can broadly find applications in industries, for the manufacture of poly-vinyl chloride and cellulose ester-based plastics like cellulose acetate and cellulose butyrate.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Dimethyl phthalate may be used as an analytical standard for the determination of the analyte in food products, vegetables, cosmetic and environmental samples by gas chromatography technique.

Other Notes

Ion Mobility: TWCCSN2 value of 134.1 Å2 [M+H]+ and 146.1 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 41320 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
525081W508500W512206
description

PESTANAL®, analytical standard

description

≥99%

description

≥99%

description

≥99%

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

application(s)

-

application(s)

flavors and fragrances

application(s)

flavors and fragrances

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

300, 400

grade

analytical standard

grade

-

grade

Fragrance grade

grade

-

assay

≥99.5% (GC)

assay

≥99%

assay

≥99%

assay

≥99%

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Dimethyl phthalate


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

294.8 °F - closed cup

flash_point_c

146.0 °C - closed cup

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters