Skip to Content
MilliporeSigma
All Photos(1)

Documents

D1200000

Dienestrol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3,4-Bis(4-hydroxyphenyl)-2,4-hexadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
Beilstein:
2053694
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dienestrol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C\C=C(c1ccc(O)cc1)\C(=C\C)c2ccc(O)cc2

InChI

1S/C18H18O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h3-12,19-20H,1-2H3/b17-3+,18-4+

InChI key

NFDFQCUYFHCNBW-SCGPFSFSSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dienestrol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Vanden Bussche et al.
Journal of chromatography. A, 1360, 229-239 (2014-08-19)
In recent years, the analysis of veterinary drugs and growth-promoting agents has shifted from target-oriented procedures, mainly based on liquid chromatography coupled to triple-quadrupole mass spectrometry (LC-QqQ-MS), towards accurate mass full scan MS (such as Time-of-Flight (ToF) and Fourier Transform
Yingying Wen et al.
Journal of chromatography. A, 1368, 18-25 (2014-12-03)
A modified Quick, Easy, Cheap, Effective, Rugged and Safe (QuEChERS) method based on the dispersive solid-phase extraction (dSPE) combined with high performance liquid chromatography (HPLC) was developed for the determination of non-steroidal estrogens in water samples. In this study, graphene
Wen-Yan Hu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 957, 7-13 (2014-03-19)
A sensitive analytical method based on packed-fiber solid-phase extraction and high performance liquid chromatography-tandem mass spectrometry (PF SPE-HPLC-MS/MS) has been developed for determination of three synthetic stilbenes in milk. The stilbenes are extracted with acetonitrile, using sodium chloride, and purified
Xiaoxia Mi et al.
Journal of chromatography. A, 1356, 54-63 (2014-07-16)
A method for analyzing 26 types of steroids in egg matrix was developed. The method used liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) in electrospray ionization mode (ESI). The procedure involved extraction with acetonitrile and removal of phospholipids by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service