Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0739000

Dextran 60/70 for performance test

Dextran 60/70 European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

biological source

bacterial (Leuconostoc mesenteroides)

grade

pharmaceutical primary standard

API family

dextran

analyte chemical class(es)

oligosaccharides

manufacturer/tradename

EDQM

application(s)

food and beverages
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextran 60/70 for performance test EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Salamon et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 186(4), 367-376 (2014-04-01)
The aim of this study was to establish co-labeling of mesenchymal stromal cells (MSC) for the detection of single MSC in-vivo by MRI and histological validation. Mouse MSC were co-labeled with fluorescent iron oxide micro-particles and carboxyfluorescein succinimidyl ester (CFSE).
Mary E Pease et al.
Investigative ophthalmology & visual science, 55(4), 2564-2573 (2014-02-22)
To determine differences in scleral permeability, as measured by diffusion of macromolecules, by using fluorescence recovery after photobleaching (FRAP), with reference to differences by mouse strain, scleral region, and the effect of experimental glaucoma. In three mouse strains (B6, CD1
Jaleh Varshosaz
Expert opinion on drug delivery, 9(5), 509-523 (2012-03-22)
Dextran is a family of natural polysaccharides that is widely under investigation for use as polymeric carriers in novel drug delivery systems. The optimal drug delivery (and consequently maximum therapeutic effect) will be accomplished when carrier systems are used mainly
Assessment of glomerular size-selective function with fractional clearance of neutral dextran.
A Remuzzi et al.
The Journal of laboratory and clinical medicine, 132(5), 360-362 (1998-11-21)
Muhammad Qasim et al.
Experimental cell research, 322(2), 277-289 (2014-02-11)
Gastrointestinal toxicity is a common adverse effect of mycophenolic acid (MPA) treatment in organ transplant patients, through poorly understood mechanisms. Phosphorylation of myosin light chain 2 (MLC2) is associated with epithelial tight junction (TJ) modulation which leads to defective epithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service