Skip to Content
MilliporeSigma

1179628

USP

Dextran 1

Dextran 1 United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

dextran

form

crystalline

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

General description

Dextran 1 is a low molecular weight dextran fraction used in hypersensitivity testing prior to dextran administration. It acts by binding circulating antibodies to reduce anaphylactic risk. The USP reference standard is used in identity and content uniformity testing.

The USP biologics carbohydrates category includes a variety of carbohydrate-based substances that are essential in the development and manufacturing of therapeutic products. These carbohydrates play crucial roles in biological processes and are often utilized as excipients, stabilizers, or active ingredients in pharmaceuticals. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of carbohydrate therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.


Still not finding the right product?

Explore all of our products under Dextran 1


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library