Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM143

Supelco

BNAs - Sandy Loam 1

pkg of 50 g

Synonym(s):

Sand matrix CRM, BNAs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM143-050

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Bis(2-ethylhexyl) phthalate
    Bis(2-chloroethoxy)methane
    4-Chloro-3-methylphenol
    2-Chloronaphthalene
    4-Chlorodiphenyl ether
    Dibenz[a,h]anthracene
    1,2-Dichlorobenzene
    1,4-Dichlorobenzene
    2,4-Dichlorophenol
    2,6-Dichlorophenol
    Diethyl phthalate
    2,4-Dimethylphenol
    Dimethyl phthalate
    Di-n-octyl phthalate
    2,4-Dinitrotoluene
    Fluoranthene
    Fluorene
    Hexachloroethane
    Hexachlorocyclopentadiene
    Isophorone
    2-Methyl-4,6-dinitrophenol
    o-Cresol
    p-Cresol
    Naphthalene
    Nitrobenzene
    2-Nitrophenol
    N-Nitrosodimethylamine
    N-Nitrosodiphenylamine
    N-Nitrosodi-n-propylamine
    Pentachlorophenol
    Phenanthrene
    Phenol
    Pyrene
    1,2,4-Trichlorobenzene
    2,4,5-Trichlorophenol
    See All (42)

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Feng Chen et al.
Marine pollution bulletin, 154, 111029-111029 (2020-04-23)
Water, suspended particulate matter (SPM), and sediment samples were collected from Salt River in Taiwan and analyzed the concentrations of 16 types of polycyclic aromatic hydrocarbons (PAHs). The analysis results were used to examine the distribution, source, partition behavior, and
Marta Pastor-Belda et al.
Talanta, 189, 543-549 (2018-08-09)
This paper presents a procedure for the determination of seven nitrophenols (NPs) in water and soil samples using stir bar sorptive extraction (SBSE) coupled to gas chromatography with mass spectrometry (GC-MS) by means of a thermal desorption unit (TDU). Microwave

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service