Skip to Content
MilliporeSigma
All Photos(1)

Documents

AP1064

Sigma-Aldrich

PhosphoDetect Anti-PDH-E1α (pSer³⁰⁰) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Pyruvate Dehydogenase pSer³⁰⁰ Rabbit pAb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer300)

Related Categories

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~44 kDa PDH-E1α protein phosphorylated at Ser300.
Recognizes the ~44 kDa PDH-E1α protein phosphorylated at Ser300 in HEK293 cells.
This PhosphoDetect Anti-PDH-E1α (pSer³⁰⁰) Rabbit pAb is validated for use in Immunoblotting, Immunocytochemistry for the detection of PDH-E1α (pSer³⁰⁰).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Ser³⁰⁰ phosphorylation site of human PDH-E1α

Application



Immunoblotting (0.5 μg/ml)
Immunocytochemistry (1 μg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 100 mM Tris-Glycine, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
HEK293 cells treated with dichloroacetate
Positive Control
HEK293 cells

Other Notes

Due to a high degree of sequence identity, this antibody is also expected to cross-react with mouse, rat, bovine, Zebra fish, and Xenopus phospho-PDH-E1α Antibody should be titrated for optimal results in individual systems.
Rardin M.J., et. al. 2009. Anal. Biochem.2, 157.
Seifert, F., et al. 2007. Biochemistry 21, 6277.
Lee, J., et al. 2007. Mol. Cell Prot. 4, 669.
Patel, M.S. and Korotchkina, L.G. 2006 Biochem. Soc. Trans.34, 217.
Korotchkina, L.G., et al. 2001. J. Biol. Chem. 40, 37223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changyeol Lee et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-11)
The fungal strain Alternaria alternata JS0515 was isolated from Vitex rotundifolia (beach vitex). Twelve secondary metabolites, including one new altenusin derivative (1), were isolated. The isolated metabolites included seven known altenusin derivatives (2-8), two isochromanones (9, 10), one perylenequinone (11)
Gretchen L Seim et al.
Nature metabolism, 1(7), 731-742 (2020-04-08)
In response to signals associated with infection or tissue damage, macrophages undergo a series of dynamic phenotypic changes. Here we show that during the response to LPS and interferon-γ stimulation, metabolic reprogramming in macrophages is also highly dynamic. Specifically, the
Yu Juan Chai et al.
Plants (Basel, Switzerland), 10(9) (2021-09-29)
Three undescribed bicyclo[3.2.1]octanoid neolignan glucosides, along with a known neolignan, were isolated from the leaves of Piper crocatum Ruiz & Pav. Their chemical structures were elucidated using extensive spectroscopic analyses including 1D and 2D NMR experiments and HR-ESI-MS analysis, as
Takeshi Yoneshiro et al.
Nature, 572(7771), 614-619 (2019-08-23)
Branched-chain amino acid (BCAA; valine, leucine and isoleucine) supplementation is often beneficial to energy expenditure; however, increased circulating levels of BCAA are linked to obesity and diabetes. The mechanisms of this paradox remain unclear. Here we report that, on cold exposure
Lifeng Yuan et al.
Cell research, 28(6), 625-643 (2018-05-29)
Cellular senescence is a fundamental cell fate playing a significant role throughout the natural aging process. However, the molecular determinants distinguishing senescence from other cell-cycle arrest states such as quiescence and post-mitotic state, and the specified mechanisms underlying cell-fate decisions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service