Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7906

Supelco

Casein from bovine milk

for use in the Total Dietary Fiber Control Kit, TDF-C10

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21

biological source

bovine milk

Quality Level

form

powder

quality

for use in the Total Dietary Fiber Control Kit, TDF-C10

technique(s)

photometry: suitable

mp

280 °C (dec.) (lit.)

suitability

suitable for use with the Total Dietary Fiber Assay Kit as internal control

UniProt accession no.

application(s)

food and beverages
general analytical

storage temp.

2-8°C

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin. However, bovine casein is not homogeneous; variants A1 and B do lead to production of β-casomorphin 7 production, while A2 does not.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Benschop et al.
Journal of clinical microbiology, 46(12), 3965-3970 (2008-10-24)
Human parechoviruses (HPeV) are widespread pathogens belonging to the Picornavirus family. Six genotypes, which have predominantly been isolated from children, are known. Data on prevalence of HPeV genotypes are generally based on cell culture, which may underestimate the prevalence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service