Skip to Content
MilliporeSigma
All Photos(1)

Documents

C6152

Sigma-Aldrich

Carmine

Dye content: minimum not established , certified by the Biological Stain Commission, powder

Synonym(s):

Alum lake of carminic acid, Cochineal, Nacarat, Natural Red 4

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
75470
EC Number:
UNSPSC Code:
12171500
NACRES:
NA.47

product name

Carmine, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content: minimum has not been established

color

red

solubility

1 mg/mL, red (2 N NH4OH)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

InChI

1S/C22H20O13/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34)

InChI key

DGQLVPJVXFOQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carmine is a red compound and is commonly used with aluminum, iron or other metal salts to enhance its activity. During smear preparation, carmine is required for chromatin staining in fresh material. In presence of aluminum chloride, it is required for the staining of glycogen.

Application

Carmine has been used for the staining of mammary glands. It has also been used for the staining of chromosomes.

Suitability

Certified for use in Orth′s lithium carbonate technique, Southgate′s mucicarmine method, and Best′s carmine for glycogen

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic background affects susceptibility to mammary hyperplasias and carcinomas in Apc(min)/+ mice.
Moser AR, et al.
Cancer Research, 61, 3480-3480 (2001)
Precocious differentiation of the virgin Wistar-Kyoto rat mammary gland.
Benton ME, et al.
Endocrinology, 140, 2659-2659 (1999)
Florence Charlot et al.
Nucleic acids research, 42(19), 11965-11978 (2014-09-28)
The eukaryotic RecA homologue Rad51 is a key factor in homologous recombination and recombinational repair. Rad51-like proteins have been identified in yeast (Rad55, Rad57 and Dmc1), plants and vertebrates (RAD51B, RAD51C, RAD51D, XRCC2, XRCC3 and DMC1). RAD51 and DMC1 are
Histophatologic Techniques null
Methods in Mammary Gland Biology and Breast Cancer Research (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service