Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-0370

Sigma-Aldrich

Schneider′s aceto-carmine

Synonym(s):

Carmine solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
grade:
SAJ special grade
form:
liquid

grade

SAJ special grade

form

liquid

availability

available only in Japan

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Suitability

nuclear staining

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service