Skip to Content
MilliporeSigma

BP458

Pyridoxine hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

5-Hydroxy-6-methyl-3,4-pyridinedimethanol hydrochloride, Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
EC Number:
200-386-2
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
3632435
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

pyridoxine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

color

white

mp

214-215 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia. Pyridoxine hydrochloride is a water soluble vitamin, which plays an important role in animal and plant metabolism.

Application

Pyridoxine hydrochloride BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Vitamins B and C Injection
  • Pyridoxine Tablets
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.


    Still not finding the right product?

    Explore all of our products under Pyridoxine hydrochloride


    pictograms

    Corrosion

    signalword

    Danger

    hcodes

    Hazard Classifications

    Eye Dam. 1

    Storage Class

    11 - Combustible Solids

    wgk

    WGK 1



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library