Skip to Content
MilliporeSigma
Pyridoxine hydrochloride British Pharmacopoeia (BP) Reference Standard

Pyridoxine hydrochloride

Pyridoxine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride, Pyridoxine hydrochloride
Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique