Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR272

Coronene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H12
CAS Number:
Molecular Weight:
300.35
Beilstein:
658468
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

525 °C (lit.)

mp

428 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1cc2ccc3ccc4ccc5ccc6ccc1c7c2c3c4c5c67

InChI

1S/C24H12/c1-2-14-5-6-16-9-11-18-12-10-17-8-7-15-4-3-13(1)19-20(14)22(16)24(18)23(17)21(15)19/h1-12H

InChI key

VPUGDVKSAQVFFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR272

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matthew R Kennedy et al.
The journal of physical chemistry. A, 116(48), 11920-11926 (2012-11-10)
π-π interactions are integral to many areas of chemistry, biochemistry, and materials science. Here we use electronic structure theory to analyze how π-π interactions change as the π-systems are curved in model complexes based on coronene and corannulene dimers. Curvature
Jing Zhang et al.
Journal of the American Society for Mass Spectrometry, 22(7), 1294-1298 (2011-09-29)
In this Application Note, we describe, for the first time, the rapid analysis of hydrophobic compounds present in environmental contaminants, which includes polycyclic aromatic hydrocarbons (PAHs) and estrogen, by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS) with the use
Hirofumi Sato et al.
Physical chemistry chemical physics : PCCP, 13(1), 309-313 (2010-11-26)
Coronene (C(24)H(12)) is a flat polyaromatic hydrocarbon consisting of seven peri-fused benzene rings and attracts lots of attention as a fragment of graphene. Using a hybrid method of quantum chemistry and statistical mechanics called RISM-SCF, which is an alternative to
Yukihiro Yoshida et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(37), 12313-12324 (2013-07-31)
Ten types of neutral charge transfer (CT) complexes of coronene (electron donor; D) were obtained with various electron acceptors (A). In addition to the reported 7,7,8,8-tetracyanoquinodimethane (TCNQ) complex of 1:1 stoichiometry with a DA-type alternating π column, TCNQ also afforded
Anna M Hiszpanski et al.
ACS nano, 7(1), 294-300 (2012-12-12)
The structuring in organic electrically active thin films critically influences the performance of devices comprising them. Controlling film structure, however, remains challenging and generally requires stringent deposition conditions or modification of the substrate. To this end, we have developed post-deposition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service