MilliporeSigma
  • Solvation structure of coronene-transition metal complex: a RISM-SCF study.
Solvation structure of coronene-transition metal complex: a RISM-SCF study.
Physical chemistry chemical physics : PCCP (2010-11-26)
Hirofumi Sato, Chisa Kikumori, Shigeyoshi Sakaki
ABSTRACT
Coronene (C(24)H(12)) is a flat polyaromatic hydrocarbon consisting of seven peri-fused benzene rings and attracts lots of attention as a fragment of graphene. Using a hybrid method of quantum chemistry and statistical mechanics called RISM-SCF, which is an alternative to QM/MM, the electronic structure and solvation structure of a coronene-transition metal complex were computed in a self-consistent manner. The binding of a ruthenium complex ([C(5)H(5)Ru](+)) was extensively studied, especially the changing of the solvation structure.
Product Description
Coronene, BCR®, certified reference material
Sigma-Aldrich
Coronene, purified by sublimation, 99%
Sigma-Aldrich
Coronene, 97%