Skip to Content
MilliporeSigma
All Photos(1)

Documents

BCR130

Quartz (particle size)

BCR®, certified reference material, 50-220 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

form

particles

manufacturer/tradename

JRC

particle size

50-220 μm

application(s)

glass & ceramic

format

neat

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR130

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service