Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2635

Supelco

Busulfan

analytical standard, for drug analysis

Synonym(s):

1,4-Butanediol dimethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2O(CH2)4OSO2CH3
CAS Number:
Molecular Weight:
246.30
Beilstein:
1791786
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

114-117 °C (lit.)

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

CS(=O)(=O)OCCCCOS(C)(=O)=O

InChI

1S/C6H14O6S2/c1-13(7,8)11-5-3-4-6-12-14(2,9)10/h3-6H2,1-2H3

InChI key

COVZYZSDYWQREU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Busulfan is a bifunctional alkylating agent, which may find application in the treatment of chronic granulocytic leukemia.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1A - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blood and Marrow Stem Cell Transplantation: Principles, Practice, and Nursing Insights (1997)
Arnon Nagler et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(28), 3549-3556 (2013-08-28)
Cyclophosphamide (Cy) combined with total-body irradiation (TBI) or with busulfan (Bu) are currently the most common myeloablative regimens used in allogeneic stem-cell transplantation (alloSCT) in adults with acute myelogenous leukemia (AML). Intravenous (IV) Bu has more predictable bioavailability and a
Yan Zhao et al.
Stem cell research & therapy, 11(1), 466-466 (2020-11-06)
Premature ovarian insufficiency (POI) is an ovarian dysfunction that seriously affects a woman's physiological health and reproduction. Mesenchymal stem cell (MSC) transplantation offers a promising treatment option for ovarian restoration in rodent POI models. However, the efficacy and mechanism of
Christopher Bredeson et al.
Blood, 122(24), 3871-3878 (2013-10-02)
We conducted a prospective cohort study testing the noninferiority of survival of ablative intravenous busulfan (IV-BU) vs ablative total body irradiation (TBI)-based regimens in myeloid malignancies. A total of 1483 patients undergoing transplantation for myeloid malignancies (IV-BU, N = 1025;
Edward A Copelan et al.
Blood, 122(24), 3863-3870 (2013-09-26)
Cyclophosphamide combined with total body irradiation (Cy/TBI) or busulfan (BuCy) are the most widely used myeloablative conditioning regimens for allotransplants. Recent data regarding their comparative effectiveness are lacking. We analyzed data from the Center for International Blood and Marrow Transplant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service