Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Please contact Customer Service for Availability | $499.00 |
About This Item
biological source
mouse
Quality Segment
assay
>98% (SDS-PAGE and HPLC)
form
liquid
specific activity
≥1.0 x 10E7 U/mg
manufacturer/tradename
Chemicon®
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 ng/μg endotoxin (of IFN-gamma; 1EU/μg)
input
sample type mesenchymal stem cell(s)
sample type hematopoietic stem cell(s)
shipped in
dry ice
storage temp.
-10 to -25°C
General description
Application
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| assay >98% (SDS-PAGE and HPLC) | assay ≥98% (SDS-PAGE) | assay >98% (SDS-PAGE and HPLC) | assay >98% (SDS-PAGE and HPLC) |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) - | technique(s) cell culture | mammalian: suitable |
| biological source mouse | biological source mouse | biological source - | biological source human |
| form liquid | form lyophilized powder | form - | form - |
| specific activity ≥1.0 x 10E7 U/mg | specific activity - | specific activity - | specific activity ≥2 x 10(E7) U/mg |
| shipped in dry ice | shipped in - | shipped in dry ice | shipped in dry ice |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



