Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0368000

Amikacin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43N5O13
CAS Number:
Molecular Weight:
585.60
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

amikacin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N([C@H]1[C@@H]([C@H]([C@@H]([C@H](C1)N)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CN)O)O[C@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)N)O)CO)C(=O)[C@@H](O)CCN

InChI

1S/C22H43N5O13/c23-2-1-8(29)20(36)27-7-3-6(25)18(39-22-16(34)15(33)13(31)9(4-24)37-22)17(35)19(7)40-21-14(32)11(26)12(30)10(5-28)38-21/h6-19,21-22,28-35H,1-5,23-26H2,(H,27,36)/t6-,7+,8-,9+,10+,11-,12+,13+,14+,15-,16+,17-,18+,19-,21+,22+/m0/s1

InChI key

LKCWBDHBTVXHDL-RMDFUYIESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amikacin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Goren et al.
The Pediatric infectious disease journal, 8(5), 278-282 (1989-05-01)
A three-drug antibiotic regimen including vancomycin and amikacin has been recommended as effective treatment in clinical settings in which Gram-positive bacteremias are a serious problem. To determine if vancomycin potentiates the tubular proteinuria associated with amikacin therapy, we studied febrile
A Bruce Montgomery et al.
Antimicrobial agents and chemotherapy, 58(7), 3714-3719 (2014-04-23)
The amikacin-fosfomycin inhalation system (AFIS) is a combination of 2 antibiotics and an in-line nebulizer delivery system that is being developed for adjunctive treatment of pneumonia caused by Gram-negative organisms in patients on mechanical ventilation. AFIS consists of a combination
Amikacin.
R D Meyer
Annals of internal medicine, 95(3), 328-332 (1981-09-01)
Sophia B Georghiou et al.
PloS one, 7(3), e33275-e33275 (2012-04-06)
Rapid molecular diagnostics for detecting multidrug-resistant and extensively drug-resistant tuberculosis (M/XDR-TB) primarily identify mutations in Mycobacterium tuberculosis (Mtb) genes associated with drug resistance. Their accuracy, however, is dependent largely on the strength of the association between a specific mutation and
P Van der Auwera
The Journal of antimicrobial chemotherapy, 27 Suppl C, 63-71 (1991-05-01)
The pharmacokinetic parameters of amikacin given once daily were compared to those of amikacin given bd at different daily dosages. In adult patients, 'peak' serum concentrations (30 min after 30 min infusion) were linearly proportional to the dose given with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service