Skip to Content
MilliporeSigma
All Photos(1)

Documents

95426

Supelco

(−)-α-Bisabolol

analytical standard

Synonym(s):

(−)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol, Levomenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥93% (GC)

optical activity

[α]/D -60±2°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC[C@](C)(O)[C@H]1CCC(C)=CC1

InChI

1S/C15H26O/c1-12(2)6-5-11-15(4,16)14-9-7-13(3)8-10-14/h6-7,14,16H,5,8-11H2,1-4H3/t14-,15+/m1/s1

InChI key

RGZSQWQPBWRIAQ-CABCVRRESA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-α-Bisabolol is a monocyclic sesquiterpene alcohol naturally found in the essential oil of different plants like Vanillosmopsis erythropappa and Matricaria chamomilla. It finds applications in clinical and cosmetic industries due to its pharmacological properties like anti-inflammatory, anti-irritant, antimicrobial and skin healing property. It also shows effective action against malignant tumor cells.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: angelica

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Insight into the apoptosis-inducing action of α-bisabolol towards malignant tumor cells: Involvement of lipid rafts and Bid
Darra E, et al.
Archives of Biochemistry and Biophysics, 476(2), 113-123 (2008)
Chiaki Nakano et al.
Chembiochem : a European journal of chemical biology, 12(15), 2271-2275 (2012-10-30)
Now found in bacteria: An increasing number of genome sequences indicate that bacteria possess a variety of terpenoid cyclase genes. The characterization of two sesquiterpene cyclase (SC) genes found in the draft genome sequence of Streptomyces citricolor is described here.
Orazio Taglialatela-Scafati et al.
Journal of natural products, 75(3), 453-458 (2012-02-09)
Apart from a large amount (ca. 2.0%) of α-bisabolol β-D-fucopyranoside (2a), the aerial parts of the Mediterranean weed Carthamus glaucus afforded an unusual triglyceride (E-2-crotonyl-1,3-distearolylglycerol, 7), two lipophilic flavonoids (6a,b), and a series of bisabolane fucopyranosides variously acylated on the
Jaekwan Kim et al.
Pharmaceutical research, 29(7), 2017-2029 (2012-03-09)
To evaluate the ability of a novel radiofrequency (RF) microporation technology based on ablation of the skin barrier to enhance topical delivery of active ingredients The influence of RF fluence and the molecular size of the absorbent on the permeation
Maryam Yousefi et al.
Natural product research, 26(2), 146-151 (2011-08-04)
The chemical composition of the essential oil obtained from the aerial parts of Betonica grandiflora Willd., growing wild in Iran, was analysed by gas chromatography-mass spectrometry for the first time. Overall, 40 volatile components were identified on the basis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service