MilliporeSigma
  • Essential oil from aerial parts of of Betonica grandiflora Willd. from Iran.
Essential oil from aerial parts of of Betonica grandiflora Willd. from Iran.
Natural product research (2011-08-04)
Maryam Yousefi, Somayyeh Gandomkar, Zohreh Habibi
ABSTRACT
The chemical composition of the essential oil obtained from the aerial parts of Betonica grandiflora Willd., growing wild in Iran, was analysed by gas chromatography-mass spectrometry for the first time. Overall, 40 volatile components were identified on the basis of their mass spectra characteristics and retention indices in which ledol (21.8%), myrtenyl acetate (21.7%), eudesm-7(11)-en-4-ol (6.5%), trans-caryophyllene (5.5%), α-bisabolol (4.9%) and isolongifolol (4.5%) were the major constituents. Oxygenated sesquiterpenes, monoterpene hydrocarbons and sesquiterpene hydrocarbons were the main groups of compounds with 45.6%, 23.1% and 18.4%, respectively. The oil was moderately active against Bacillus subtilis and Staphylococcus aureus and inactive against Enterococcus faecalis and Klebsiella pneumoniae.
Sigma-Aldrich
(−)-α-Bisabolol, ≥93% (GC)
(−)-α-Bisabolol, primary reference standard
Supelco
(−)-α-Bisabolol, analytical standard