Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

94426

Supelco

Enoxacin sesquihydrate

analytical standard

Synonym(s):

1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-naphthyridine-3-carboxylic acid sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17FN4O3 · 1.5H2O
CAS Number:
Molecular Weight:
347.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤15 wt. % water

solubility

1 M NaOH: 50 mg/mL, clear, colorless to faintly yellow

suitability

conforms to structure for Proton NMR spectrum

application(s)

clinical testing

format

neat

SMILES string

O.O.O.CCN1C=C(C(O)=O)C(=O)c2cc(F)c(nc12)N3CCNCC3.CCN4C=C(C(O)=O)C(=O)c5cc(F)c(nc45)N6CCNCC6

InChI

1S/2C15H17FN4O3.3H2O/c2*1-2-19-8-10(15(22)23)12(21)9-7-11(16)14(18-13(9)19)20-5-3-17-4-6-20;;;/h2*7-8,17H,2-6H2,1H3,(H,22,23);3*1H2

InChI key

DKNNITGJCMPHKE-UHFFFAOYSA-N

Application

Enoxacin sesquihydrate may be used as a reference standard for the determination of enoxacin in plasma by high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous assay of fluoroquinolones and theophylline in plasma by high-performance liquid chromatography.
Davis JD, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 621(1), 105- 109 (1993)
Anna Margaret McDonnell et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-25)
Cervical cancer is a major cause of death in females worldwide. While survival rates have historically improved, there remains a continuous need to identify novel molecules that are effective against this disease. Here, we show that enoxacin, a drug most
Julieta Griboff et al.
Food chemistry, 332, 127380-127380 (2020-07-01)
The occurrence of 46 antibiotics (amphenicols, cephalosporins, dihydrofolate reductase inhibitors, fluroquinolones, macrolides, nitrofurans, penicillins, quinolones, sulfamides and tetracyclines) in Argentinean market fish were investigated by UPLC-MS/MS. Veterinary and human antimicrobials enrofloxacin, clarithromycin, roxithromycin, doxycycline and oxytetracycline were detected in 100%

Articles

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service