Skip to Content
MilliporeSigma

Key Documents

Skip To

32982

Ciprofloxacin-d8 hydrochloride hydrate

VETRANAL®, analytical standard

Synonym(s):

1-Cyclopropyl-6-fluoro-4-oxo-7-piperazin-d8-1-yl-1,4-dihydro-quinoline-3-carboxylic acid hydrochloride hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17D8H10FN3O3 · HCl · xH2O
CAS Number:
Molecular Weight:
375.85 (anhydrous basis)
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+8

SMILES string

O.Cl.[2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)C4CC4)C1([2H])[2H]

InChI

1S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19H,1-6H2,(H,23,24);1H;1H2/i3D2,4D2,5D2,6D2;;

InChI key

ARPUHYJMCVWYCZ-CORRJWLCSA-N

General description

Ciprofloxacins are classified under the fluoroquinolones group of antibiotics.

Application

Ciprofloxacin-d8 hydrochloride hydrate may be used to prepare ciprofloxacin-d8 that can be used as an internal standard for the determination of ciprofloxacin in:
  • Groundwater samples collected from cemetery areas, urban wastewater samples and dairy products by ultra-high performance liquid chromatography-triple quadrupole mass spectrometry (UHPLC-MS/MS) equipped with electrospray ionization (ESI) source and multiple reaction monitoring (MRM) mode of detection.
  • Hospital wastewater and chicken feces by LC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
3389911343131134346
description

VETRANAL®, analytical standard

description

VETRANAL®, analytical standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

technique(s)

gas chromatography (GC): suitable, HPLC: suitable

technique(s)

gas chromatography (GC): suitable, HPLC: suitable

technique(s)

-

technique(s)

-

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

application(s)

clinical testing

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

grade

analytical standard

grade

analytical standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters